LaCrosse Glass Rain Gauge
# de catalogue 470309-888
Fournisseur: LA CROSSE TECHNOLOGY
Spécifications
- Description:Rain gauge
- Cat. no.:470309-888
Spécifications
A propos de cet article
Clear glass rain gauge measures 5" (12cm) of rainfall and can be mounted either onto a post or placed into the ground. Large inch digits and clear measuring lines make it easy to read and removable glass tube for easy cleaning.
- Measures rainfall in inches/centimeters
- Mount can be secured to post or staked into ground
- Aligned with GLOBE protocols
Customers Who Bought This Also Bought