ContactUsLinkComponent
Waterproof ExStik® II Dissolved Oxygen Meter Kit
Waterproof ExStik® II Dissolved Oxygen Meter Kit
# de catalogue 470310-230
Fournisseur:  NEW GENIE GROUP INC SE
Kit Do Meter Exstik Ii 1.4X6.9X1.6In
Waterproof ExStik® II Dissolved Oxygen Meter Kit
# de catalogue 470310-230
Fournisseur:  NEW GENIE GROUP INC SE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Dissolved Oxygen Meter Kit
  • Cat. no.:
    470310-230
Spécifications
A propos de cet article
The DO600-K is a convenient kit which adds versatility for measuring dissolved oxygen in laboratories or out in the field with a 16' (5 m) extension cable and probe weight guard for taking easy measurements in tanks or streams.
  • Oxygen level displayed as % Saturation or Concentration (mg/L [ppm])
  • Adjustable Altitude Compensation (0 - 20000 feet in 1,000 feet increments)
  • Adjustable Salinity Compensation from 0 to 50 ppt
  • Analog bargraph indicates trends
  • Memory stores up to 25 data sets with DO and Temperature reading
  • Self calibration on power up
Delivery: Supplied complete with ExStik® II DO meter, DO electrode, protective sensor cap, DO603 replacement membrane Kit, 16' (5 m) extension cable, weighted probe guard, four 3V CR-2032 batteries, 48" (1.2 m) neck strap and hard carrying case.
Customers Who Bought This Also Bought