I-100, Valve Unit, Buchi
# de catalogue 76266-304
Fournisseur: Buchi
Spécifications
- Description:Vacuum Controller I-100 Valve unit with 4" Display
- Cat. no.:76266-304
Spécifications
A propos de cet article
Vacuum Controller I-100 with Valve Unit for house or 3rd party vacuum source.
- Functional design fulfills essential needs of vacuum regulation
- User-friendly and intuitive handling thanks to self-explaining user interface
- Relevant information is displayed at a glance
The I-100 is an easy to use interface that controls the house or 3rd party vacuum sources digitally through the controller and valve unit.
Customers Who Bought This Also Bought