Avantor® Hichrom, Guard Cartridge Holder for HPLC Columns
# de catalogue 75830-798
Fournisseur: HICHROM LIMITED
Spécifications
- Description:All-Guard Cartridge Holder for 2.1 - 4.6 mm I.Ø Columns
- Cat. no.:75830-798
- Supplier No.:80101/N
Spécifications
A propos de cet article
This guard cartridge holder promotes increased column lifetime.
- Improved hardware
- Higher pressure rating
- Low internal swept volume
This guard cartridge holder is supplied in new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.
Customers Who Bought This Also Bought