R-100 Rotavapor® Rotary Evaporator System, Büchi
Catalog # 76266-274
Supplier: Buchi
Specifications
- Description:Rotavapor® Rotary Evaporator System, SR-100, V P+G, F-105
- Cat. no.:76266-274
- Electrical:110 V
- Condenser Type:Vertical Coil/Chiller
- Joint Size:24/40 NS
- Coating:P+G
Specifications
About this item
The R-100 Rotavapor® is an entry level Rotavapor®, meeting the most essential needs in evaporation. The V-100 vacuum pump and F-105 recirculating chiller is included and controlled by the I-100 Interface controller.
- Economical
- Solid
- Expandable
- Reliable operation
- Flexibility
The R-100 Rotavapor® has a user-friendly manual-lift mechanism for easy positioning of the evaporation flask in to and out of the water bath. The V-100 vacuum pump and F-105 recirculating chiller is included and is controlled by the I-100 Interface controller. This full system has all of the components needed for efficient evaporation. The patented Combi-clip allows for easy flask handling and vapor duct removal.
Certifications: CE and CSA compliant.
Ordering information: All accessories and connections are included for out of the box usage. Optional safety shield, 1 l evaporation flasks, different joint size glassware, valves for in-house cooling and vacuum are available as accessories (sold separately). Please contact your Avantor representative for more information.
Frequently Bought Together