Avantor® Hichrom, Spanner Wrenches
Fournisseur: HICHROM LIMITED
Certificats
Certificats
A propos de cet article
Analytical guard holder wrench set contains a pair of stainless steel wrenches.
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$46.59 to $465.92
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$133.66
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$131.41 to $136.31