Porcelainware Starter Kit, United Scientific Supplies
# de catalogue 76293-324
Fournisseur: United Scientific Supplies
Spécifications
- Description:Porcelainware Starter Kit
- Cat. no.:76293-324
Spécifications
A propos de cet article
A customized kit that includes top-selling porcelain products.
- High temperature resistance porcelain
This small porcelainware set includes the most commonly used items.
Delivery: Kit includes Crucible, Porcelain, Low Form, 50ml; Crucible, Porcelain, High Form, 50ml; Casserole, Porcelain, 200ml; Mortar and Pestle, 275ml; Evaporating Dish, 80ml; Clay Pipe Triangle, 2"; Crucible Tongs, Nickel Plated, Steel, 9"
Customers Who Bought This Also Bought