Ladies 38" Knot Button iPad® Labcoats, White Swan/Meta
Fournisseur: White Swan
Certificats
A propos de cet article
These labcoats feature five pockets (two lower patch pockets, a breast pocket with pen stitch, an inside cell and inside iPad® pocket).
- Made of 100% easy care cotton twill
- Hand access slits
The iPad ® pocket comfortably and securely holds an iPad® in the labcoat. Sewn down back belt with accent darts.
Spécifications
- Catalog No:
- 10024-628
- 10024-632
- 10024-644
- 10024-642
- 10024-646
- 10024-624
- 10024-616
- 10024-638
- 10024-636
- 10024-620
- 10024-618
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill
- 100% easy care cotton twill