Laboratory Jacks, Painted Aluminum and Stainless Steel, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Laboratory support jacks are constructed of painted aluminum and stainless steel. The scissor jack design enables height adjustment with a simple turn of the knob.
- Aluminum and stainless steel
- Easy to control height knob
4×4" platform that can be vertically adjusted from a closed position of 2.5" up to 6". Maximum load capacity is 10 kg.
6×6" platform that can be vertically adjusted from a closed position of 3" up to 10.5". Maximum load capacity is 20 kg.
8×8" platform that can be vertically adjusted from a closed position of 3" up to 11.75". Maximum load capacity is 25 kg.
12×12" platform that can be vertically adjusted from a closed position of 4" up to 19.75". Maximum load capacity is 45 kg.
Customers Who Bought This Also Bought