Laboratory Jack, Anodized Aluminum and Stainless Steel, United Scientific Supplies
# de catalogue 76293-612
Fournisseur: United Scientific Supplies
Spécifications
- Cat. no.:76293-612
- Maximum Load:20 kg
- Platform Size:4.5×5.5"
Spécifications
A propos de cet article
Laboratory support jack is constructed of chemically resistant anodized aluminum and stainless steel.
- Chemically resistant
- Stainless steel and aluminum construction
The scissor jack design enables height adjustment with a simple turn of the knob. Sturdy design can support loads up to 20 kg. Platform height can be vertically adjusted from closed position of 2.75" up to 10.5".
Customers Who Bought This Also Bought