Chemistry Hardware Assortment, United Scientific Supplies
# de catalogue 76293-318
Fournisseur: United Scientific Supplies
Spécifications
- Description:Chemistry Hardware Assortment
- Cat. no.:76293-318
Spécifications
A propos de cet article
An assortment of hardware most often used in general chemistry labs.
- Durable support stands
- Strong metal construction
When performing experiments often times you need to hold an item securely. This set provides a variety of equipment to hold beakers, boiling flasks, or test tubes safety.
Delivery: Kit includes: (1) Beaker Tongs; (1) Burette Clamp; (1) Burner Tripod, 3" ring, 9" legs; (1) Mohr's Pinchcock Clamp; (1) Clamp Holder, right angle; (1) Clay Pipe Triangle, 2"; (1) Crucible Tongs, nickel-plated steel, 9"; (1) Ring Support with Clamp, 4"; (1) Support Stand with Rod, 5×8"/24" rod; (1) 6-Place Plastic Test Tube Rack; (1) Test Tube Clamp; (1) Wire Gauze with Ceramic Center, 5×5".
Customers Who Bought This Also Bought