Volumetric Glassware Starter Kit, United Scientific Supplies
# de catalogue 76293-316
Fournisseur: United Scientific Supplies
Spécifications
- Description:Volumetric Glassware Starter Kit
- Cat. no.:76293-316
Spécifications
A propos de cet article
A customized volumetric glassware assortment designed for general laboratory use.
- Graduated cylinders are double scale
- Volumetric flasks have snap caps
Measuring the volume of solutions in your lab is a keystone for experiments. This kit provides a wide range of volumetric measuring devices based on your specific need.
Delivery: Kit includes: (6) Measuring Pipettes, Class B, 10 ml with 0.10 ml Graduations; (6) Transfer Pipettes, 25 ml; (1) Double Scale Cylinder, 50 ml; (1) Double Scale Cylinder, 100ml; (1) PTFE stopcock Straight Bore Burrette, 50 ml; (1) Filtering Flask, 500ml; (1) Snap Cap Volumetric Flask, 50 ml; (1) Snap Cap Volumetric Flask, 100 ml; (1) Separatory Funnel, 500 ml.
Customers Who Bought This Also Bought