Rubber Filter Aid, United Scientific Supplies
Catalogue # 76293-286
Fournisseur: United Scientific Supplies
Spécifications
- Description:Rubber Filter Aid
- Cat. no.:76293-286
Spécifications
À propos de cet article
This molded rubber filter disc is designed to support and seal in place a filter crucible or a funnel in the neck of a filter flask.
- Molded rubber
- Flat bottom forms a tight seal
Inside diameter of the socket at top is 32mm, and outside diameter of the disc is 76 mm. The bottom of the disc is flat so that it seals tightly against the lip of the flask.