Stamped Steel Heavy Support Stands with Rods, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Heavy stamped steel bases with black enamel finish feature an added steel plate on the back, which makes them about 50% heavier than regular stamped steel bases.
- Zinc plated rod
The steel plate also facilitates a sturdier threading of the rod into the base.
Ordering information: Bases and Rods are also available individually.
Delivery: Includes a removable, zinc-plated steel rod.
Customers Who Bought This Also Bought