Cast Iron Support Stands with Rods, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Tip-free stability for experiments.
- Glassware can sit directly on base
- Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
Customers Who Bought This Also Bought