Stamped Steel Two-Hole Support Stands with Rods, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Tip-free stability for experiments.
- Features two tapped holes
- zinc plated rod
Stamped steel bases with black enamel finish features 2 tapped holes. Hole at center of the shorter side is for standard ring stand use. Hole at center of the longer side is for applications such as titrations, specific gravity determinations and more.
Ordering information: Bases and Rods are also available individually.
Delivery: Includes a removable, zinc-plated steel rod.
Customers Who Bought This Also Bought