Support Stand and Ring Sets, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Each set contains a stamped steel base and a zinc-plated steel rod, along with 2, 3, 4, or 5 cast iron rings with clamps.
- Sets comes with 2, 3, 4 or 5 rings of varied diameters
- Base is stamped steel and the plate underneath is made of steel
- Base weighs 2 lbs.
The support stand will support up to 10 lbs.
Customers Who Bought This Also Bought