WebSeal™ 96-Well Plates, Barcoded, for Vanquish™ UHPLC Systems, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
WebSeal™ PP well plates for Vanquish™ UHPLC systems, provides advanced sample security and sample integrity.
- SBS format and barcode enables automatic inventory in the vanquish autosampler
- Easy inventory management through barcode reading with empty shelf-level detection and well plate verification
Monitor and manage an inventory of up to 9 deep well plates or 20 shallow well plates (15 mm height) in the vanquish charger with an additional three positions in the Vanquish autosampler, for a maximum of 8832 samples.
The barcodes indicate a well plate identification and the correct side orientation of the plate to ensure the correct position in the vanquish autosampler and charger unit when loading and running the analytical sequence. Plates cannot be accepted in the wrong orientation.
Customers Who Bought This Also Bought