Plastic Clamps for Jointed Glassware
Fournisseur: United Scientific Supplies
New Product
Certificats
A propos de cet article
Hold jointed sockets securely.
- Manufactured from polypropylene
- Outstanding chemical resistance
- Maximum operating temperature of 135 °C
- Autoclavable
Plastic conical joint clips are designed to securely hold joint size glassware together. The clamps are non-corrosive, and will not scratch glassware.
Customers Who Bought This Also Bought