Cork Borer, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Sturdy, high-quality plated brass cork borers with solid brass handles, except for the set of three which includes plastic handles.
- Plated brass
- Solid metal handles
Use this set to make custom sized holds in cork stoppers. To be used only on cork stoppers.
Delivery: Bore sizes by set are as follows:
Set of 3 includes: 4 mm, 6.5 mm, 8 mm
Set of 6 includes: 4 mm, 5 mm, 6.5 mm, 8 mm, 9.5 mm, 11 mm
Set of 9 includes: 4 mm, 5 mm, 6.5 mm, 8 mm, 9.5 mm, 11 mm, 12.75 mm, 16 mm, 17.5 mm
Set of 12 includes: 4 mm, 5 mm, 6.5 mm, 8 mm, 9.5 mm, 11 mm, 12.75 mm, 14 mm, 16 mm, 17.5 mm, 20 mm
Set of 3 includes: 4 mm, 6.5 mm, 8 mm
Set of 6 includes: 4 mm, 5 mm, 6.5 mm, 8 mm, 9.5 mm, 11 mm
Set of 9 includes: 4 mm, 5 mm, 6.5 mm, 8 mm, 9.5 mm, 11 mm, 12.75 mm, 16 mm, 17.5 mm
Set of 12 includes: 4 mm, 5 mm, 6.5 mm, 8 mm, 9.5 mm, 11 mm, 12.75 mm, 14 mm, 16 mm, 17.5 mm, 20 mm
Customers Who Bought This Also Bought