Scott® Paper Towels
Catalog # 76243-557
Supplier: Kimberly-Clark
Specifications
- Ø:1.5" (roll)
- Width:8.0"
- Color:White
- Description:Scott® paper towels
- Cat. no.:76243-557
- Packaging:12 rolls/case
- Roll Length:425'
Specifications
About this item
Scott essential paper towel rolls give soft feel with the premium performance of commercial paper towels.
- Compatible with a variety of Kimberly-Clark paper towel dispensers
Absorbency pockets absorb lots of water fast for superior hand drying performance.
Certifications: ECOLOGO Forest Stewardship Council (FSC).
Customers Who Bought This Also Bought