VWR® Open End Serological Pipettes
Wide, open end design is ideal for viscous and high particulate solutions, as well as slurries such as Sephadex.
- Temperature range: –20 to 50 °C (–4 to 122 °F)
- Pipettes are latex-free
- Sterilized by irradiation
- DNase-/RNase-free, non-pyrogenic, and non-cytotoxic
- Graduated in 1/10 ml
- Pipettes are not autoclavable
- Dispenser box for easy opening
Shelf life: 3 years after month of production.
Graduations are calibrated for accurate dispensing (within ±2%). Negative graduations up to –0.3 ml (total volume: 1.3 ml).
All pipettes are printed with a batch number for traceability. Meets BSE/TSE requirements.
Certifications: Manufactured in a class 100000 cleanroom environment. Manufactured under EN ISO13485 and ISO9001 quality management systems.
Packaging: Pipettes are available individually packed (in peel-to-open wraps with lot number for quality traceability), or bulk packed in easy-to-open bags.
Customers Who Bought This Also Bought