ContactUsLinkComponent
ADC® Adscope® 605 Infant Clinician Stethoscopes
ADC® Adscope® 605 Infant Clinician Stethoscopes
# de catalogue 470309-768
Stethoscope Adscope 605 Black 30.5In
ADC® Adscope® 605 Infant Clinician Stethoscopes
# de catalogue 470309-768
Spécifications
  • Color:
    Black
  • Description:
    Adscope 605
  • Cat. no.:
    470309-768
Spécifications
A propos de cet article
Ultra-sensitive diaphragm.
  • Combination infant chest piece
  • Includes ID tag
  • Non-chill bell and diaphragm retaining ring
The ADC® Adscope® 605 offers legendary acoustics, performance, and functionality for the smallest patients. The combination infant chest piece offers both an ultra-sensitive diaphragm, 1.125" in diameter, for greater amplification and crisper high-frequency transmission and a 0.875" diameter bell for unsurpassed low-frequency response. The clinician headset, designed to withstand thousands of cycles, features a double-leaf tension spring and a reinforcing yoke molded into flexible 22" PVC tubing.
Customers Who Bought This Also Bought