Infinity Plan Dry Objective 100X, LW SCIENTIFIC SE | Avantor
Infinity Plan Dry Objective 100X, LW SCIENTIFIC SE
100X Infinity Plan Dry
Catalog # 470309-550
Supplier:   LW Scientific
Fournisseur #:  MSO-100X-IPDR
Infinity Plan Dry Objective 100X, LW SCIENTIFIC SE
Catalog # 470309-550
Supplier:  LW Scientific
Infinity Plan Dry Objective 100X, LW SCIENTIFIC SE
Infinity Plan Dry Objective 100X, LW SCIENTIFIC SE
100X Infinity Plan Dry
Price
$995.15
Log in pour la disponibilité
Each
Product Details & Documents
No oil required.
  • Directly fits most microscopes
  • No oil mess
  • Increases speed of slide scanning
This new 100× DRY Infinity PLAN objective will bring a radical change in the world of microscopy. With clarity, resolution, and flat-field-of-view that rivals the best 100× OIL Infinity PLAN objectives. Why use oil? Imagine how quickly doctors, techs, and students can scan cytology slides without ever slowing down; plus the satisfaction and relief after so many years of struggling with oil.
This objective installs onto any Boreal, LW Scientific microscope (Mi5, i4, Innovation, and Revelation lll) and can also be installed onto other microscope brands like Olympus, Unico, Motic, Labomed, Swift, Accuscope, and Nikon (Nikon e200 requires an adaptor ring. Call for more information).
Spécifications
  • Désignation:
    100X Infinity Plan Dry