TLC Tank Rack, PTFE-Coated Aluminum, Chemglass
# de catalogue 80060-620
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:TLC tank rack, PTFE-coated aluminum
- Cat. no.:80060-620
- Supplier No.:CG-1185-20
Spécifications
A propos de cet article
PTFE-coated aluminum construction.
- Designed to fit TLC developing tanks
- Rack holds six 20 cm×20 cm×1/8" or twelve 20 cm×20 cm×1/16" plates
Customers Who Bought This Also Bought