TLC Developing Tanks, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Heavy-duty, molded TLC developing tank.
- For use with plates up to 20.3×20.3 cm (8×8")
- Interior tank dimensions: 27×7×25.4 cm (W×D×H) (10.625×2.875×10")
The weight of the tank is approximately 15 lbs., and the bottom is ground to ensure stability.
Delivery: Tank is supplied complete with lid (replacement lids also available individually)
Customers Who Bought This Also Bought