Tefzel® Flangeless Ferrules, Chemglass
# de catalogue 80060-478
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Tefzel® Flangeless Ferrule
- Size:1/8"
- Cat. no.:80060-478
- Supplier No.:CG-1164-F-02
Spécifications
A propos de cet article
Ferrules are designed to form a zero dead-volume connection.
- Use with flangeless nut to ensure tubing is held firmly in place
- Yellow finish
Ferrule is placed on the end of the tubing with the cone pointing toward the nut, flat end towards the PTFE fitting.
Customers Who Bought This Also Bought