Cryo FreezeBox and Accessories, Crystal Technology
Fournisseur: CRYSTAL TECHNOLOGY & INDUSTRIES
Certificats
A propos de cet article
Cooling equipment for your samples. Different Ice Cores available to create your own FreezeBox.
- No need for dry ice
- Less contamination
- Easy to use and bench space friendly
Simple to use. Store the Freezebox Ice Core in ultra-low temperature and place desired module inside. Work on your bench easily with no need for dry ice.
Ordering information: Accessories not included.
Customers Who Bought This Also Bought