Claisen Adapters, with [ST] Outer Joints, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapters feature outer standard taper joints on both top arms and lower inner joint.
- Overall dimensions as listed
- Joint sizes as listed
All three joints on each adapter are the same size.
Customers Who Bought This Also Bought