Jacketed Distillation Adapter, Chemglass
# de catalogue 80066-182
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Jacketed Distillation Adapter, 75°
- Cat. no.:80066-182
- ST Joint Size:24/40 - 10/30
- Supplier No.:CG-1075-01
Spécifications
A propos de cet article
Adapter is vacuum jacketed and can be used as a distilling head to connect a condenser with the top of the distillation column.
The 10/30 top outer joint is designed for use with 75 mm immersion thermometers. Inner joints are at a 75° angle to one another and are of same size.
Customers Who Bought This Also Bought