Kjeldahl Distilling Trap Adapters, Varied [ST] Joints, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam.
Adapters with joints of non-similar sizes at the upper and lower portion of the adapter.
Customers Who Bought This Also Bought