Kjeldahl Distilling Trap Adapters, Same Size [ST] Joints, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam.
With top standard taper outer and lower inner joints, both being the same size.
Customers Who Bought This Also Bought