Distillation Adapters, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapters have a #7 Chem-Thread at the top for a vacuum-tight seal of plain stem thermometers.
Adapters feature the same size inner joints on the bottom and sidearm at a 75° angle to one another.
Delivery: Supplied with a compression cap and Viton O-Ring
Customers Who Bought This Also Bought