Distillation Adapters, with Tooled Rubber Fitting, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapters have a tubulation at the top for use with rubber fitting.
Fitting permits insertion of plain stem, partial immersion thermometers. With the same size inner joints on the bottom and sidearm at a 75° angle to one another.
Delivery: Supplied with one rubber fitting.
Customers Who Bought This Also Bought