ContactUsLinkComponent
Airfree® Schlenk Solvent Transfer Manifolds, Chemglass
Airfree® Schlenk Solvent Transfer Manifolds, Chemglass
# de catalogue 80064-968
Fournisseur:  Chemglass
Small Business Enterprise
SOLVENT TRANSFER MANIFOLD, AIRFREE
Airfree® Schlenk Solvent Transfer Manifolds, Chemglass
# de catalogue 80064-968
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Description:
    Airfree® Schlenk Solvent Transfer Manifold
  • Cat. no.:
    80064-968
  • Supplier No.:
    AF-0558-01
Spécifications
A propos de cet article
Designed to purify and transfer solvents under inert conditions.
  • Equipped with a Chem-Vac™ Chem-Cap®
  • With 24/40 joint
Manifold permits transfer of solvents directly into an evacuated storage vessel with 24/40 standard taper outer joints. A 0 to 4 mm Chem-Vac™ Chem-Cap® valve is used for vacuum control.
Customers Who Bought This Also Bought