Tefzel® Flangeless Tubing Nuts, Chemglass
# de catalogue 80060-480
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Tefzel® Flangeless Nut, 1/8"
- Cat. no.:80060-480
- Supplier No.:CG-1164-N-02
Spécifications
A propos de cet article
Flangeless nut eliminates the need to flange tubing.
- Use with a flangeless ferrule for a leak-free connection of FEP tubing
- Nut is molded out of Tefzel®
- Nut has a 1/4"–28 thread
Simply insert tubing through the nut, attach the ferrule (flat end towards the PTFE fitting) to the end of the tubing, and finger tighten the nut into place. Nut has a 1/4"–28 thread, and is molded out of natural Tefzel®, a soft fluoropolymer resistant to chemical attack.
Customers Who Bought This Also Bought