Solvent Reservoir Bottles, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Solvent reservoir bottles, glass bottle only.
- Plastic-coated flat bottom
- Blue tinted
Bottles are blue tinted, with a plastic-coated flat bottom. The coating provides additional safety by containing the glass fragments from accidents and allowing enough time to transfer samples or discarding safely. For complete assembled solvent reservoir systems, see CG-1167-11, CG-1167-12, CG-1167-12, or CG-1167-14 series.
Customers Who Bought This Also Bought