Modified Claisen Adapters, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Claisen-style adapters with an additional standard taper outer joint at a 45° angle to the vertical outer joint.
Customers Who Bought This Also Bought