Heating Mantles, Upper Portion for Unjacketed Cylindrical Vessels, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Glass fabric heating mantle with a lengthwise slit, held together with a hook and lace closure.
- For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels
- 115 volts
Delivery: Supplied complete with a separable 4', 2-wire cord and locking connector.
Customers Who Bought This Also Bought