Septa for Target 10-425 Glass Screw-Thread Vials, Thermo Scientific
Fournisseur: Avantor
Certificats
A propos de cet article
Thermo Scientific™ 10 mm septa are designed for use with 10-425 threaded caps.
Customers Who Bought This Also Bought
Thermo Scientific™ 10 mm septa are designed for use with 10-425 threaded caps.