ContactUsLinkComponent
First Aid Only Trauma Pads, Acme United
First Aid Only Trauma Pads, Acme United
# de catalogue 76287-296
Fournisseur:  First Aid Only
Trauma Pad Sterile 5In X 9In 4/ REFILL
First Aid Only Trauma Pads, Acme United
# de catalogue 76287-296
Fournisseur:  First Aid Only
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Trauma Pads
  • Size:
    5×9"
  • Cat. no.:
    76287-296
  • Packaging:
    4 per Refill
Spécifications
A propos de cet article
Highly absorbent dressings are designed to stop bleeding.
  • Sterile
  • Provides padding and protection
  • Designed to wick away fluids and disperse them evenly
Sterile, absorbent trauma dressings help stop the bleeding associated with deep lacerations, abrasions, burns, penetration wounds and fractures. These highly absorbent dressings provide padding and protection for large wounds. Each quick-wicking pad features a moisture-resistant barrier to ensure superior performance.
Customers Who Bought This Also Bought