Distillation Adapter, with Two [ST] Joints, Drip Tip, and Hose Connection, Chemglass
# de catalogue 80066-172
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Distillation Adapter, 100 mm Center to Center
- Cat. no.:80066-172
- ST Joint Size:14/20
- Supplier No.:CG-1072-B-14
Spécifications
A propos de cet article
With two standard taper inner joints, parallel, at a 75° angle to the connecting tube.
- 14/20 Joint
- 100 mm center to center
- 75° angle to the connecting tube
Adapter has a sealed-in drip tip and serrated hose connection for vacuum source.
Customers Who Bought This Also Bought