Distillation Adapters, with Two [ST] Joints, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
With two standard taper inner joints, parallel, and at a 75° angle to the connecting tube.
Customers Who Bought This Also Bought
With two standard taper inner joints, parallel, and at a 75° angle to the connecting tube.