Distillation Adapters, with Thermometer Joint, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapters have a standard taper outer joint at the top of the main tube for use with jointed thermometers.
- With inner joints on the bottom and sidearm at a 75° angle to one another
- For use with the corresponding size jointed thermometers
- Manufactured by Chemglass
The 10/18 size is used with 10/18, 25 mm immersion thermometers while the 10/30 is for use with 10/30, 75 mm immersion thermometers.
Customers Who Bought This Also Bought