ContactUsLinkComponent
First Aid Only™ Cotton Tipped Applicators, Acme United
First Aid Only™ Cotton Tipped Applicators, Acme United
# de catalogue 76287-318
Fournisseur:  First Aid Only
Cotton Tipped Applicator 3In 100/VIAL
First Aid Only™ Cotton Tipped Applicators, Acme United
# de catalogue 76287-318
Fournisseur:  First Aid Only
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    3"
  • Description:
    Cotton Tip Applicators
  • Cat. no.:
    76287-318
  • Packaging:
    100 per Vial
  • Handle Type:
    Wooden
Spécifications
A propos de cet article
Cotton tipped swabs are ideal for cleansing, for removal of debris, and for applying topical medications.
  • Features a soft cotton tip that is absorbent and will not lint
  • Stored in an easy to handle vial
  • Wooden shafts are odorless, tasteless, and splinter-free
Swabs are packaged in an easy to handle vial so that it is small enough to be stored anywhere.
Customers Who Bought This Also Bought