Luer Adapters, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapter allows for 1/8" O.D. PTFE tubing to be connected to glassware via a standard taper joint.
- Can be used with standard luer lock stainless steel needles
- Supplied with CG-1055-20 female luer
- Useful with vacuum manifolds or any other air sensitive transfer applications
Adapters can also be used with standard luer lock stainless steel needles (see CG-3075 series). Adapters are supplied with a CG-1055-20 female luer to 1/4"–28 PEEK fitting. Ferrules, nuts, and tubing must be ordered separately.
Customers Who Bought This Also Bought