Universal Inlet Adapters, PTFE, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Unique adapter is made entirely of PTFE and is supplied complete with a series of five PTFE ferrule-style cones.
- Inside diameters run from 3.5-6.5 mm
- Color-coded cones for easy identification
- Replacement PTFE cones are available in sets of 5 or individually
These cones have inside diameters that run from 3.5 to 6.5 mm and seal against the inserted tube by means of a threaded compression screw. Cones are color-coded for easy identification.
Customers Who Bought This Also Bought