EPA Vial Replacement Septa, Ace Glass Incorporated
# de catalogue 89064-066
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Ø:24/400
- Description:PTFE-faced silicone septa
- Cat. no.:89064-066
- Supplier No.:8781-45