arium® UV Lamp (185/254 nm), Sartorius Corp.
Catalog # 75803-118
Supplier: Sartorius
Specifications
- Description:arium® mini UV Lamp (185 | 254 nm) for arium® mini, arium® mini essential and arium® mini plus
- Cat. no.:75803-118
- Supplier No.:H2O-CEL1
Specifications
About this item
Ultrapure water free of TOC.
- Horizontal installation; optimized temperature gradient
- Effective breakdown of organic compounds
- Prevents the growth of microorganisms
- Easy replacement
- TOC for purified water* <5 ppb
Horizontally aligned, the UV lamp delivers especially reliable results. Unlike vertical units, the temperature gradient is less pronounced and therefore does not affect the activity of UV radiation. With two different wavelengths, the UV lamp reliably removes total organic compounds (TOCs), efficiently preventing microbial growth. At 185 nm, organic compounds are oxidized and at 254 nm, microorganisms are killed off.
Customers Who Bought This Also Bought