Septa for Target DP™ 9-425 Screw-Thread Vial Caps, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Thermo Scientific™ 9 mm septa are available in PTFE/red rubber, PTFE/silicone or red PTFE/white silicone/red PTFE to suit a variety of applications.
Customers Who Bought This Also Bought